Imprimer
Disposal canisters, with screw cap
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2012 2823 2824 2013
216-7313EA 55.6 EUR
216-7313 SEMA2823 SEMA2824 SEMA2013
Disposal canisters, with screw cap
Flacons Jerricans
HDPE, coloured
  • Good chemical resistance
  • Good temperature resistance
  • With a hollow handle, filling opening and vent screw
  • Stackable
For collecting chemicals.