Imprimer
Disposable syringes, 3-parts, ONCE
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1959 1961
612-0320EA 68.2 EUR
612-0320 SEMA1961
Disposable syringes, 3-parts, ONCE
Seringues Seringues médicales
These syringes have precise pistons. The piston and seal are made of silicone rubber.
  • Gas sterilised and non-pyrogenic
Sealing, sterility and depyrogenation are subject to continuous controls.
Certifications: Conforms with ISO 7886.
Cdt: Individually wrapped.