Imprimer
Strips of 12 PCR 0,2 ml tubes or domed PCR cap strips, SoftStrips®
Fournisseur: BIOZYM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
711060 711061 711062 711063 711064 711066 711068 711070 711071 711072 711073 711074 711076 711078
BZYM711060EA 168 EUR
BZYM711060 BZYM711061 BZYM711062 BZYM711063 BZYM711064 BZYM711066 BZYM711068 BZYM711070 BZYM711071 BZYM711072 BZYM711073 BZYM711074 BZYM711076 BZYM711078
Strips of 12 PCR 0,2 ml tubes or domed PCR cap strips, SoftStrips®
Tubes Tubes PCR
Tubes without caps. Cap strips must be ordered separately.
  • High Profile
  • DNA-, DNase-, RNase-free