Imprimer
Microtome rotatif, HM 325
Fournisseur: Epredia
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
902100 902100ER 902100A
720-2704EA 0 EUR
720-2704 SHAN902100ER SHAN902100A
Microtome rotatif, HM 325
Microtomes et groupes de microtomes Microtomes
Cut high-quality paraffin sections without a power supply with the HM 325. It features retraction at the return travel to protect the specimen and manual coarse advance for fast working.
  • One-handed zero orientation head
  • Quick trim button
  • Section counter
  • Guide rail guidance for the knife holder
  • Large, easy-to-clean waste tray extends below the blade holder
Informations de livraison: Please contact your local Avantor office for details of knives, blades and a full range of accessories.
New Product
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
L×P×H 420×490×280 mm
Poids 23 kg