Imprimer
Disposable syringes, 3-piece
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1950 2955
612-0607EA 160 EUR
612-0607 SEMA2955
Disposable syringes, 3-piece
Seringues Seringues médicales
The piston and its seal are made of silicone rubber.
  • Available with centred and eccentric tip
Certifications: Conforms with ISO 7886, 2955, 5993, 10521.