Imprimer
Laboratory trays, HDPE
Fournisseur: SEMADENI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1317 1316
215-0235EA 57.2 EUR
215-0235 216-7348
Laboratory trays, HDPE
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
Laboratory trays made of white HDPE.
  • Stackable
  • Good chemical resistance
  • Good temperature resistance
With reinforcing ribs and supporting edge.