Imprimer
Dog serum
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S2900-100 S2900-010 S2900-050
S2900-100EA 0 EUR
S2900-100 S2900-010 S2900-050
Dog serum
Sérums
Dog serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Dog serum is used as a blocking agent in immunoassays and controls.