Imprimer
Calf serum
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S0400-100 S0400-500 S040R-500 S040H-100 S040H-500 S0400-1000 S040J-500
S0400-100EA 19.9 EUR
S0400-100 S0400-500 S040R-500 S040H-100 S040H-500 S0400-1000 S040J-500
Calf serum
Sérums
Calf serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Calf serum is used as a blocking agent in immunoassays and controls.