Imprimer
Newborn calf serum
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S0750-500 S0750-100 S075H-100 S075H-500 S075G-500
S0750-500EA 109 EUR
S0750-500 S0750-100 S075H-100 S075H-500 S075G-500
Newborn calf serum
Sérums
Newborn Calf Serum (NBCS) originates from 3 to 10 day old calves. It is a high quality, cost effective replacement for Foetal Bovine Serum (FBS) and contains a somewhat higher proportion of proteins and immunoglobulin, but fewer growth factors. With proper cell adaptation, it has been shown to perform very well in cell culture applications where routine cell growth and cell culture maintenance is the objective.