Imprimer
Human plasma
Marques : Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S4180-500 S4180-100
S4180-500EA 475 EUR
S4180-500 S4180-100
Human plasma
Plasma
Human plasma is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.
Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).
Attention: Products of human origin should be considered potentially infectious and handled accordingly.