Imprimer
Donkey serum
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S2170-100 S2170-500
S2170-100EA 78.9 EUR
S2170-100 S2170-500
Donkey serum
Sérums
Donkey serum may be used as a blocking agent and controls in immunoassays. For immunohistochemistry and immunocytochemistry, donkey serum was used between 5 to 20% as a blocking agent in various studies.