Imprimer
Sheep serum
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S2350-100 S2350-500 S235H-500 S2350-5000
S2350-100EA 27.1 EUR
S2350-100 S2350-500 S235H-500 S2350-5000
Sheep serum
Sérums
For immunohistochemistry applications, sheep serum is frequently used as a blocking solution. Using sheep serum is useful for blocking the background cross reactivity with immunoglobulins which are already present in the tissue. When used before application of the primary antibody, sheep serum can eliminate Fc receptor binding of the primary and secondary antibody.