Imprimer
Rabbit serum
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S2500-100 S2500-500 S250H-500
S2500-100EA 20.6 EUR
S2500-100 S2500-500 S250H-500
Rabbit serum
Sérums
Rabbit serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Rabbit serum is used as a blocking agent in immunoassays and controls.