Imprimer
Blotting papers, Owl™
Fournisseur: THERMO OWL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FP-1 FP-2 FP-3 FP-4 FP-5 FP-6 FP-7
700-2190EA 173 EUR
700-2190 OWLAFP-2 OWLAFP-3 700-2191 700-2192 OWLAFP-6 OWLAFP-7
Blotting papers, Owl™
Papiers buvard
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.