Imprimer
Sporicide, Contec® ProChlor
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FBT102PC FBC502PC FBC102PC
141-0883EA 244 EUR
141-0883 141-0882 CONTFBC102PC
Sporicide, Contec® ProChlor
Produits nettoyants Désinfectants
ProChlor est un biocide sporicide à action rapide innovant conçu pour une utilisation dans les salles blanches pharmaceutiques. À base d'acide hypochloreux.
  • Efficacious against bacteria, fungi, moulds, yeasts, viruses and spores
  • Log 6 kill against Bacillus subtilis in 60 s saves time spent on biodecontamination
  • Filtered to 0,2 µm in a Grade B cleanroom ensures the product is free from contamination and particulates
  • Contains no quaternary ammonium or surfactant so it's very low residue
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom
Contec ProChlor is 0,2 µm filtered, and filled in a Grade A environment. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Informations de commande: Sterile version available for Grade A and B cleanrooms
Informations de livraison: Supplied as either trigger sprays for small areas or 1 L and 5 L capped containers for when larger volumes are required.
Attention: Use biocides safely. Always read the label and product information before use.
No hazard classification means only basic PPE required and no special disposal is required