Imprimer
Wide opening vials, Virtuoso™
Fournisseur: CHROMACOL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60180-VT300 60180-VT301 60180-VT302
548-1941EA 43.2 EUR
548-1941 548-1942 548-1943
Wide opening vials, Virtuoso™
Fioles
These wide opening screw neck, crimp top and snap top vials are designed exclusively for use with the Virtuoso™ vial identification system. They are made of superior quality 33 expansion borosilicate clear Type 1, Class A.
  • Virtuoso™ ID patch, specific for the Virtuoso™ vial identification system
  • Same high performance level as with other Thermo Scientific chromatography vials
  • Sample data is printed directly on the vial's V-patch
Virtuoso™ enables you to record the information your laboratory needs directly on the vial. The patches provide a surface that allows direct printing that is resistant to all common laboratory solvents.