Imprimer
Erlenmeyer flasks, baffled, with membrane screw cap
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1142/12 1142/08 1142/10 1142/14
214-0358EA 207 EUR
214-0358 214-0428 214-0429 214-0430
Erlenmeyer flasks, baffled, with membrane screw cap
Ballons Flacons Erlenmeyers
PYREX®, borosilicate glass.
  • Four equidistant baffles provide consistent agitation when used with compatible orbital shakers
  • Excellent chemical resistance, high temperature resistance
  • Flasks can be autoclaved with the membrane screw cap fastened
Ideal for aerobic microbiology and cell culture. Complete with blue polypropylene GL 45 screw cap with a gas-permeable PTFE membrane that permits gaseous exchange whilst contents remain sterile.