Imprimer
Bottles, narrow neck, square, HDPE, with screw caps, LaboPlast®
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0331-0050 0331-0020 0331-0100
BURK0331-0050EA 1 EUR
BURK0331-0050 BURK0331-0020 BURK0331-0100
Bottles, narrow neck, square, HDPE, with screw caps, LaboPlast®
Flacons Flacons en plastique
HDPE, translucent.
  • Shatterproof
  • Special labelling area for labels or manual lettering
  • Leakproof
Square bottle for packing and storing of liquids, powders and granules.