Imprimer
Vacuum blood collection tubes, without additives, Vacutest®
Fournisseur: KIMA SAS DI CHIARIN RENZO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
149430 149415 149425
720-2695EA 395 EUR
720-2695 720-2693 720-2694
Vacuum blood collection tubes, without additives, Vacutest®
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
PET, transparent.
  • Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls
  • PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected
  • Labels are made from self-adhesive paper for specific medical use, size 40x20 mm, printed in three colours
  • Available in different sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
The closure system of the test tubes is composed of a pierceable butyl rubber stopper and a polyethylene safety cap. The stopper has a hollow in the piercing spot to protect operator from possible contact. The safety cap protecting against the aerosol and splashes prevents the operator from coming into contact with the pierceable rubber stopper and allows for easy recapping of the test tube.
18 months shelf-life.