Imprimer
VWR® Standard Line, Bottles, Wide Neck, PET, Round or Square, with Screw Cap
Fournisseur: LAB3 OWN PRODUCTION
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
331-0155EA 112 EUR
331-0155 331-0161 331-0165 331-0167
VWR® Standard Line, Bottles, Wide Neck, PET, Round or Square, with Screw Cap
Flacons Flacons en plastique
PET, clear.
  • 34 mm neck for all bottles
  • Tamper-evident screw caps
  • Manufactured from food grade PET
For basic sample collecting. Non sterile, lightweight, round or square. All caps are HDPE.
Attention: Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904.