Imprimer
Industrial food scales, Valor® 2000
Fournisseur: OHAUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30072346 30072347 30072316 30072318 30072317 30072349 30072315 30072348
611-4392EA 326 EUR
611-4392 611-4393 611-4389 611-4391 611-4390 611-4395 611-4388 611-4394
Industrial food scales, Valor® 2000
Balances and Scales
Waterproof compact scales designed for the food industry. Features include keypad with two mechanical switches on V22PWE models and four on V22XWE models, adjustable feet, level bubble and cover with
spring to protect power connector of the scale from ingress of moisture, especially when the scale is washed down. Dual LED display back and front enable two operators to work with the Valor® 2000 at the same time. Choice of scale with plastic or stainless steel housing, the models with stainless steel housing also have a check weighing application mode.
  • Stabilisation times of 0,5 seconds
  • Flow Thru™ design protects scales from water damage - water drains away even after submersion
  • Thick polycarbonate keypad is impact and puncture resistant
  • Easy to clean and sanitary design support HACCP certified systems
  • Carrying handle and rechargeable lead acid battery with up to 50 hours of continuous usage time, allowing for portable operation