Imprimer
Safety shoes, lace-up, Steelite™ WORK Arx Safety Trainer FW33
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FW33BKR40 FW33BKR38 FW33BKR39 FW33BKR41 FW33BKR45 FW33BKR46 FW33BKR42 FW33BKR43 FW33BKR44 FW33BKR37 FW33BKR47 FW33BKR48 FW33BKR36
PRTWFW33BKR40EA 56.2 EUR
PRTWFW33BKR40 PRTWFW33BKR38 PRTWFW33BKR39 PRTWFW33BKR41 PRTWFW33BKR45 PRTWFW33BKR46 PRTWFW33BKR42 PRTWFW33BKR43 PRTWFW33BKR44 PRTWFW33BKR37 PRTWFW33BKR47 PRTWFW33BKR48 PRTWFW33BKR36
Safety shoes, lace-up, Steelite™ WORK Arx Safety Trainer FW33
Chaussures et bottes
Sporty low cut safety trainer with split leather upper and a dual-density EVA/rubber sole resistant to oil and heat (300 °C).
  • Shock absorber heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Ideal for light industrial work.
Colour: Black or White.
Certifications: EN ISO 20345:2011; S1P, SRC, HRO