Imprimer
Cold traps, long version, type KFL
Fournisseur: KGW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17380 17382 17381 17362
KGWA17380EA 1220 EUR
KGWA17380 KGWA17382 KGWA17381 KGWA17362
Cold traps, long version, type KFL
Pièges à vide
These cold traps (cold finger condensers) are constructed in the same way as standard cold traps.
  • Available with or without spouts
  • Complete versions include Dewar flask and plastic ring
The condensation area of the cold traps has been extended in order to trap more condensation. In addition to this, the complete cold trap versions also have a larger plastic ring and a bigger Dewar flask.
An upgraded version is a long version cold trap with an outlet spout. These cold traps are constructed in the same way as standard cold traps but have a GL 32 glass thread together with a screw-on cap, which is the actual spout. With the help of this spout the condensate can easily be poured out of the cold trap. Therefore the cold trap is easily to clean.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585 (DURAN made by Schott).