Imprimer
Foetal bovine serum (FBS), premium grade
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S181B-100 S181B-500 S181BH-100 S181BH-500
S181B-100EA 86.3 EUR
S181B-100 S181B-500 S181BH-100 S181BH-500
Foetal bovine serum (FBS), premium grade
Sérums
Sterile filtered, South American origin foetal bovine serum (FBS) is derived from clotted whole blood aseptically collected from foetus via cardiac puncture. Each manufactured batch is rigorously controlled, from the collection of serum through to final packaging.
  • Pre-selected and derived from own collection sources
  • No time consuming batch testing required
  • Defined endotoxin level: <5 EU/ml
  • Defined haemoglobin level: <25 mg/100 ml
  • Guaranteed growth promotion >80%*
  • Low batch-to-batch variation
*tested on SP2/O-Ag14, HELA, L929 and MRC-5 cells.
This premium grade product is intended for 'off-the-shelf' use. No batch-specific reservations will be held.
Certifications: Biowest is a certified ISO 9001:2008 company. Registered by the French Ministry of Agriculture (EC regulation No. 1069/2009) under agreement no. FR 49.231.001 for the production of animal sera.