Imprimer
Moulded pathology media vials, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
128139S 128044F
LSUK128139SEA 98.8 EUR
LSUK128139S LSUK128044F
Moulded pathology media vials, WHEATON®
Fioles
Manufactured from neutral Type 1B glass. These vials are suitable for all pharmaceutical, media, diagnostic and sample collection applications.
  • Thick walled moulded glass, chemically inert and with excellent thermal properties
  • Aluminium caps fitted with non-inhibitory rubber liners for storage of cultures and allows cell growth
  • Self sealing PP caps provide leak proof seal