Imprimer
Safety boots, lace-up, Compositelite™ WORK Thor FC12
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FC12BKR39 FC12BKR40 FC12BKR41 FC12BKR42 FC12BKR43 FC12BKR44 FC12BKR45 FC12BKR46 FC12BKR38 FC12BKR48 FC12BKR47
PRTWFC12BKR39EA 83.2 EUR
PRTWFC12BKR39 PRTWFC12BKR40 PRTWFC12BKR41 PRTWFC12BKR42 PRTWFC12BKR43 PRTWFC12BKR44 PRTWFC12BKR45 PRTWFC12BKR46 PRTWFC12BKR38 PRTWFC12BKR48 PRTWFC12BKR47
Safety boots, lace-up, Compositelite™ WORK Thor FC12
Chaussures et bottes
Metal-free, anti-static, lightweight boot made from water resistant nubuck fabric and a durable PU/rubber sole, resistant to oil and heat (200 °C).
  • Fur lining
  • Shock-absorbent heel
  • Composite toecap (200 J)
  • Puncture resistant composite midsole
Ideal for cold conditions.
Colour: Black
Certifications: EN 20345:2011+A1:2007; S3, HRO, SRC.