Imprimer
Single channel pipettes, mechanical, variable volume, VITLAB®
Fournisseur: VITLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1641012 1641008 1641010 1641000 1641002 1641004 1641006
VITL1641012EA 366 EUR
VITL1641012 VITL1641008 VITL1641010 VITL1641000 VITL1641002 VITL1641004 VITL1641006
Single channel pipettes, mechanical, variable volume, VITLAB®
Pipettes Pipetteurs
These piston-operated manual pipettes are ideal for demanding laboratory applications.
  • Central, large pipetting button for easy, uniform piston movement
  • Large, ergonomic eject button for rapid replacement of the tips
  • Simple volume adjustment for right- and left-handers
  • Precise, 4-position volume indicator, with integrated zoom function for optimal readability
  • Colour-coded for easy selection of the right pipette tip
  • Ergonomic finger loop for convenient handling
  • Integrated calibration function for easy calibration without the need for tools
  • Corrosion-resistant pistons and ejectors for long product life
  • Completely autoclavable at 121 °C
DE-M marked
Informations de livraison: Supplied with silicone oil resp. grease (Size 5 and 10 ml), sample bag with pipette tips, quality certificate, and instruction manual