Imprimer
Colonnes capillaires, phases traditionnelles, SLB-IL100, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28884-U 28886-U 28887-U
28884-UEA 870 EUR
28884-U 28886-U 28887-U
Colonnes capillaires, phases traditionnelles, SLB-IL100, Supelco®
Colonnes pour chromatographie
These are ionic liquid GC columns. They offer a great choice for analysis of neutral and polarisable (contain double and/or triple C-C bonds) analytes.
  • Non-bonded 1,9-di(3-vinylimidazolium)nonane bis(trifluoromethylsulfonyl)imide phase
  • Resistant to damage from moisture/oxygen
  • High thermal stability - maximum temperatures up to 230 °C