Imprimer
Inlet base seals
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
290GA083 290GA084 290GA091 290GA092 290GA094 290GA122 290GA082 290GA081
555-9130EA 102 EUR
555-9130 555-9223 555-9222 555-9221 555-9219 555-9215 555-9037 555-9038
Inlet base seals
Scellés Dispositifs mécaniques de scellage
Ideal for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenol.
Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports.