Imprimer
Chaussures de travail, sabots, X-Light, 711130/711135
Fournisseur: Abeba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
711130/35 711130/36 711130/37 711130/38 711130/39 711130/40 711130/41 711130/42 711130/43 711130/44 711130/45 711130/46 711130/47 711130/48 711135/35 711135/36 711135/37 711135/38 711135/39 711135/40 711135/41 711135/42 711135/43 711135/44 711135/45 711135/46 711135/47 711135/48
ABEB711130/35EA 87.1 EUR
ABEB711130/35 ABEB711130/36 ABEB711130/37 ABEB711130/38 ABEB711130/39 ABEB711130/40 ABEB711130/41 ABEB711130/42 ABEB711130/43 ABEB711130/44 ABEB711130/45 ABEB711130/46 ABEB711130/47 ABEB711130/48 ABEB711135/35 ABEB711135/36 ABEB711135/37 ABEB711135/38 ABEB711135/39 ABEB711135/40 ABEB711135/41 ABEB711135/42 ABEB711135/43 ABEB711135/44 ABEB711135/45 ABEB711135/46 ABEB711135/47 ABEB711135/48
Chaussures de travail, sabots, X-Light, 711130/711135
Chaussures et bottes
Anti-static safety shoe made from smooth leather upper and a lightweight, fuel resistant PU sole.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable active comfort insole provides optimised comfort, orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Lightweight PU sole with high slip resistance and shock-absorbing system
  • Non foldable, adjustable heel strap
  • Instep area with elastic band
Suitable for use in kitchens and in food processing industries.
Colour: White or black
Certifications: DGUV 112-191; EN ISO 20345:2011; OB, SRC.