Imprimer
Sharps disposal container Multi-Safe euroMatic®
Fournisseur: SARSTEDT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
77.3897.040 77.3897.070
SARS77.3897.040EA 576 EUR
SARS77.3897.040 SARS77.3897.070
Sharps disposal container Multi-Safe euroMatic®
Poubelles Collecteurs de déchets tranchants
PP, yellow/red.
  • With innovative day lid and final closure
  • Single-handed use of day lid in daily routine: at the push of the bottom, the day lid opens automatically
With universal removal aids (also ideal for scalpels) for disposal of all common cannulas, disposable syringes, blood pipettes and scalpels.
Diameter lid: 173 mm