Imprimer
Dropping funnels, cylindrical, without pressure balance
Fournisseur: SMITH SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8SD1/21T 8SD1/22T
SMIT8SD1/21TEA 75.1 EUR
SMIT8SD1/21T SMIT8SD1/22T
Dropping funnels, cylindrical, without pressure balance
Entonnoirs Ampoules de coulée
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape with straight discharge tube, socket and plastic stopper.