Imprimer
Rubans adhésifs pour salles propres
Fournisseur: Texwipe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TPA2024WH TPA1048CL TPA1048WH
817-0287EA 1700 EUR
817-0287 817-0271 817-0277
Rubans adhésifs pour salles propres
Rubans adhésifs
The TPA series is constructed from a low density polyethylene (LDPE) backing with an acrylic adhesive. This is low in halogens, leachable chlorides and heavy metals, providing users with the ideal cleanroom tape solution. TPA cleanroom tape is ideal for colour coding and product identification, and its high conformability makes it well suited for sealing and packaging materials in critical environments.
  • Acrylic adhesive
  • Latex-free
  • High conformability, adhesion and cohesion
  • Packaged in reusable press and seal cleanroom bags
Length: 32,9 m
Different colours available.
Cdt: The tapes are uniquely packaged in press and seal cleanroom bags providing additional convenience for end users.