Imprimer
PCR strip tube plates, 96-well, VersiPlate
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB-1800 AB-1805
732-2589EA 160 EUR
732-2589 732-2591
PCR strip tube plates, 96-well, VersiPlate
Microplaques Plaques PCR
VersiPlates comprise strips of eight tubes linked to each other forming the familiar 12×8 or 96-well ANSI/SBS format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
  • Easier sample handling as multiple strips of samples can be handled simultaneously
  • Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
  • Can be used in instruments with or without a VersiFrame
  • Reusable PC frame can serve as a
En savoir plus