Imprimer
Dissection boards, disposable, DispoCut™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M630-1 M630-2 M630-3
SIMPM630-1EA 489 EUR
SIMPM630-1 SIMPM630-2 SIMPM630-3
Dissection boards, disposable, DispoCut™
Planches de dissection
These dissection boards are made from a copolymer resin in order to increase impact and temperature performance. The board is strong yet inexpensive enough to throw away. It is especially developed to provide a clean, safer and more efficient way of handling infectious tissue specimens for the pathologist.
  • Can be used on both sides
  • Printed with helpful imperial and metric dimensional scales in inches, centimetres and millimetres
  • Chemically inert, withstands most oils, solvents and water (at regular temperatures)
  • Surface will not dull knifes
  • Perfect for use with dissecting pins
Each surface has many conveniently located rulers including seven dimensional circles from 4 to 16 mm in diameter and five larger ones from 2 to 6 cm in diameter. All sizes of DispoCut™ dissecting boards are ideal for pathology labs, medical schools, classroom dissections, forensic labs, medical research labs, pharmaceutical labs, surgery and more.
Sterilisation: DispoCut™ may be wiped down with alcohol with no effect on the material. DispoCut™ is being autoclaved (in excess of 3 years) in many situations. There are some extreme settings which will be damaging and we suggest a trial to determine performance in your equipment. Ethylene oxide gas sterilisation has no effect of any kind on DispoCut™ and may be repeated any number of times.
Temperature performance range: −27...+71 °C
Melting point: 162 °C