Imprimer
Cryoboxes
Fournisseur: NEOLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2-2911 2-2912
NEOL2-2911EA 0 EUR
NEOL2-2911 NEOL2-2912
Cryoboxes
Boîtes Boîtes pour cryotubes
Cryoboxes are made from PP and comes with an alphanumerically labelled grid for identifying contents.
  • Designed for use in deep freezers at temperature of –90 to +120 °C
  • Transparent overlapping turned-up lid
Informations de livraison: Assorted colour pack contains one of each colour blue, green, pink, yellow, transparent and orange.