Imprimer
Nitrile gloves, Peha-soft® Nitrile Fino
Fournisseur: HARTMANN PAUL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9421974 9421984 9421994
HART9421974EA 17.7 EUR
HART9421974 HART9421984 HART9421994
Nitrile gloves, Peha-soft® Nitrile Fino
Gants
These disposable gloves are made from very soft nitrile. Ideal for use in all areas with contamination risks.
  • Inner coating for easier donning
  • Tear-resistant, excellent fit and dexterity
  • Textured fingertips for improved grip
  • Latex-free and therefore ideal for people with type I allergies
Suitable for applications involving the handling of chemicals and disinfectants.
Certifications: EN 455, EN 374; Cat. I+II
Food contact approved