Imprimer
Reaction vessel bubblers, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
GB1/29 GB1/19 GB2/19SC GB2/24SC GB2/29SC
201-0770EA 188 EUR
201-0770 BIBBGB1/19 BIBBGB2/19SC BIBBGB2/24SC BIBBGB2/29SC
Reaction vessel bubblers, Quickfit®
Barboteurs
Pyrex® borosilicate glass.
  • Side arm to facilitate filling with mineral oil
  • With gas inlet to provide continuous flow of inert gas (N₂ or Ar) over the reaction
  • Bubbler with inlet complete with detachable plastic hose connection to allow easy and safe attachment to 9 mm flexible tubing
Ideal for closing reaction mixture from the atmosphere. Bubblers have capacity of approximately 5 ml.