Imprimer
Glove liners, 265R
Fournisseur: Showa
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
265-M 265-S 265-XL 265-L
112-5752EA 68.7 EUR
112-5752 112-5753 112-5754 112-5751
Glove liners, 265R
Gants
These seamless knitted nylon liners with thin nitrile coating protect the hand from oils, hydrocarbons, grease and abrasion.
  • Light, elastic, low-lint glove, resistant to deformation
  • Breathable back reduces perspiration
  • Anatomical shape: Designed for easy movement and continuous wear
  • Elasticated cuffs which protect the wrist
Ideal for people working in agriculture, horticulture and construction.
Length: 240 - 270 mm
EN 388 (3131); Cat. II
Certifications: EN 388 (3131X); Cat. II.