Imprimer
Capillary columns, TraceGOLD™ TG-5SilMS Fast
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26096-5780 26096-2760
555-8646EA 884 EUR
555-8646 555-8653
Capillary columns, TraceGOLD™ TG-5SilMS Fast
Colonnes pour chromatographie
These columns are designed for low bleed and reduced susceptibility to oxidation.
  • Low polarity, silarylene phase
  • High thermal stability - maximum temperatures up to 330/350 °C