Imprimer
Fingertight adapters, Color-Sleeve
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-55084-5 JR-55084-10 JR-55083-5 JR-55083-10 JR-55082-5 JR-55082-10 JR-55081-10 JR-55080-5 JR-55080-10 JR-55081-5 JR-55085-5 JR-55089-10 JR-55091-5 JR-55091-10 JR-55090-5 JR-55090-10 JR-5508X-24 JR-55089-5 JR-55088-5 JR-55088-10 JR-55087-5 JR-55087-10 JR-55086-5 JR-55086-10 JR-5508X-12 JR-55085-10
JOURJR-55084-5EA 15.5 EUR
JOURJR-55084-5 JOURJR-55084-10 JOURJR-55083-5 JOURJR-55083-10 JOURJR-55082-5 JOURJR-55082-10 JOURJR-55081-10 JOURJR-55080-5 JOURJR-55080-10 JOURJR-55081-5 JOURJR-55085-5 JOURJR-55089-10 JOURJR-55091-5 JOURJR-55091-10 JOURJR-55090-5 JOURJR-55090-10 JOURJR-5508X-24 JOURJR-55089-5 JOURJR-55088-5 JOURJR-55088-10 JOURJR-55087-5 JOURJR-55087-10 JOURJR-55086-5 JOURJR-55086-10 JOURJR-5508X-12 JOURJR-55085-10
Fingertight adapters, Color-Sleeve
Raccords et connecteurs
These PP adapters are used for colour coding different solvent lines. They turn ¹/₄" hex-head nuts into fingertight fittings.
  • Designed for PPS fittings for ¹/₁₆" tubing, low and high pressure
  • Available in twelve colours for easy identification
Informations de commande: Assorted pack JR-5508X-24 includes all 12 colours and JR-5508X-12 includes blue, red, green, yellow, black, white adapters.