Imprimer
Tubing, PTFE, Omnifit
Fournisseur: OMNIFIT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
008T16-030-20 008T16-030-200 008T16-050-20 008T16-050-200 008T16-080-20 008T16-080-200 008T16-100-20 008T16-100-200 008T32-150-10 008T32-150-100
OMNI008T16-030-20EA 217 EUR
OMNI008T16-030-20 OMNI008T16-030-200 OMNI008T16-050-20 OMNI008T16-050-200 OMNI008T16-080-20 OMNI008T16-080-200 OMNI008T16-100-20 OMNI008T16-100-200 OMNI008T32-150-10 OMNI008T32-150-100
Tubing, PTFE, Omnifit
Tuyaux Transfer Tubing
PTFE, white.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible
Excellent resistance to acids, bases and solvents.