Imprimer
Sample loops for Cheminert® valves, stainless steel
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-SLC-86-500 JR-SLC-86-50 JR-SLC-86-5 JR-SLC-86-250 JR-SLC-86-20 JR-SLC-86-1000 JR-SLC-86-100 JR-SLC-86-10
JOURJR-SLC-86-500EA 149 EUR
JOURJR-SLC-86-500 JOURJR-SLC-86-50 JOURJR-SLC-86-5 JOURJR-SLC-86-250 JOURJR-SLC-86-20 JOURJR-SLC-86-1000 JOURJR-SLC-86-100 JOURJR-SLC-86-10
Sample loops for Cheminert® valves, stainless steel
Matériel de chromatographie Chromatographie, Boucles d'échantillonnage
These VICI Jour sample loops are made of high quality stainless steel 316.
  • Fit all Cheminert® C1, C2, C2V, and C3 type valves
  • High pressure resistance, low permeability
  • Tolerance of ±0,05 mm and pressure rating of < 350 bar
They are cut using an electrolytic cutting process, cleaned with ultrasound and high pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.
Informations de livraison: Supplied with stainless hex-head nuts and stainless ferrules for reliable connections.