Imprimer
Sample loops for Valco valves
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-SLV-75-10 JR-SLV-75-100 JR-SLV-75-1000 JR-SLV-75-250 JR-SLV-75-5 JR-SLV-75-50 JR-SLV-75-500 JR-SLV-75-20
JOURJR-SLV-75-10EA 115 EUR
JOURJR-SLV-75-10 JOURJR-SLV-75-100 JOURJR-SLV-75-1000 JOURJR-SLV-75-250 JOURJR-SLV-75-5 JOURJR-SLV-75-50 JOURJR-SLV-75-500 JOURJR-SLV-75-20
Sample loops for Valco valves
Matériel de chromatographie Chromatographie, Boucles d'échantillonnage
These sample loops are made from PEEK tubing and fittings. Mechanically strong, chemically inert polymer ideal for HPLC applications. Softener-free polyethylene caps are used to avoid contamination of the tubing.
  • Fits Valco W and UW type valves
  • Tolerance of ±0,025 mm
PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.
Informations de livraison: Supplied with PEEK hex-head nuts and PEEK ferrules.