Imprimer
picoLUCENT™ PLUS-HRP Kit
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-09 786-002 786-09T 786-424 786-264 786-165
786-09EA 261 EUR
786-09 786-002 786-09T GENO786-424 GENO786-264 GENO786-165
picoLUCENT™ PLUS-HRP Kit
Réactifs d'électrophorèse Western Blotting Reagents
The picoLUCENT™ PLUS-HRP kit is based on an ultra sensitive luminol substrate that produces chemiluminescence upon reaction with horseradish peroxidase. The chemiluminescence light emission can be recorded by a short exposure to autoradiography films.
  • Low picogram detection (10⁻¹²) of antigens
  • Low noise (signal/background ratio)
Informations de livraison: Complete kit includes a non animal protein blocking agent (NAP-BLOCKER™) and wash buffer (femto-TBST™) and contains sufficient reagents for 25 mini blots (8×7,5 cm) or 1,500 cm² of PVDF or nitrocellulose membrane.