Imprimer
Microplaques et boîtes de culture Multidish pour culture cellulaire, Nunclon™ Sphera™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174929 174925 174927 174930 174931 174932
392-0377EA 1230 EUR
392-0377 392-0607 392-0608 734-2767 734-2768 734-2769
Microplaques et boîtes de culture Multidish pour culture cellulaire, Nunclon™ Sphera™
Microplaques Microplaques pour culture cellulaire
The Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment. The surface supports many different cell types and their ability to generate spheroids.
  • Optimises spheroid formation
  • Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
  • Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimises variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth, providing a better model system for studying tumour cell progression and efficacy of anticancer agents in vitro.
Nunclon™ Sphera™ surface demonstrates superior quality for embryoid body (EB) formation of pluripotent stem cells with minimal spontaneous differentiation. The resultant cells are able to differentiate into all three germ layers.