Imprimer
PYREX® Bottles, Media-lab, Clear, with High Temp Cap and Pouring Ring, DWK Life Sciences
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1516/01RED 1516/02RED 1516/04RED 1516/08RED 1516/10RED 1516/12RED 1516/06RED
BIBB1516/01REDEA 125 EUR
BIBB1516/01RED BIBB1516/02RED BIBB1516/04RED BIBB1516/08RED BIBB1516/10RED BIBB1516/12RED BIBB1516/06RED
PYREX® Bottles, Media-lab, Clear, with High Temp Cap and Pouring Ring, DWK Life Sciences
Flacons Flacons en verre
The screw caps on these bottles are manufactured from a temperature-resistant thermoplastic polymer, allowing them to be hot air sterilised up to 180 °C, compared to the 140 °C for bottles with a polypropylene screw cap.
  • Printed with trace code for downloadable batch certificate
  • Withstands hot air sterilisation up to 180 °C
  • PTFE cap liner resists chemical attack
  • White graduations and marking spot
Bottles are manufactured from temperature-resistant and chemically-resistant borosilicate glass. All bottles are supplied complete with thermoplastic polybutylene terephthalate (PBT) cap, PTFE cap liner, and pouring ring.
The 25 ml size bottle features a glass lip and does not require a pouring ring.
Certifications:
Complies with ISO 4796.