Imprimer
Microtubes à centrifuger
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
409111/2 409111/6
DELT409111/2EA 109 EUR
DELT409111/2 DELT409111/6
Microtubes à centrifuger
Tubes Microtubes à centrifuger
Made of transparent PP.
  • Certified RNAse, DNAse and PCR inhibitors free
  • Withstand centrifugation at 20000×g
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for use in liquid nitrogen, autoclave and for boiling applications, and can be used at temperatures down to −190 °C. Products ideal for long term sample storage.
Informations de commande: Screw caps must be ordered seperately.