Imprimer
Crimp top headspace vials
Fournisseur: THERMO NATIONAL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C4020-S20 C4020-S10
NSCAC4020-S20EA 233 EUR
NSCAC4020-S20 NSCAC4020-S10
Crimp top headspace vials
Fioles
These headspace vials are manufactured to provide uniform glass thickness which insures even heat distribution for consistent sampling reliability. Use Tin plate seals with magnetic pick autosamplers. Pressure release seals are designed to open when internal pressures exceed 3,0 ±0,5 bar, protecting equipment during analysis of highly volatile samples.