Imprimer
Dialysis tubing, SERVAPOR®
Fournisseur: Serva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44139.01 44139.02 44144.01 44144.02 44145.01 44145.04 44146.01 44146.04 44148.01 44148.02
SERA44139.01EA 0 EUR
SERA44139.01 SERA44139.02 SERA44144.01 SERA44144.02 SERA44145.01 SERA44145.04 SERA44146.01 SERA44146.04 SERA44148.01 SERA44148.02
Dialysis tubing, SERVAPOR®
Tuyaux Membranes de dialyse
Dialysis tubing is made of cellulose membrane. It contains low amounts of impurities such as sulphides and heavy metals.
  • High chemical and physical resistance
  • pH range: 2 to 12
  • Temperature range: 4 to 60 °C